1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%
1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%
1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%
Thermo Scientific Chemicals

1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%

CAS: 203303-01-9 | C9H6F14O2 | 412.123 g/mol
Quantity:
5 g
25 g
Catalog number L16599.06
also known as L16599-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS203303-01-9
IUPAC Name2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol
Molecular FormulaC9H6F14O2
InChI KeyXQULMKMNFZLURS-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (GC)≥88.0%
Melting Point104-114°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only