1-Chloro-3-fluoro-2-propanol, 95%
1-Chloro-3-fluoro-2-propanol, 95%
1-Chloro-3-fluoro-2-propanol, 95%
Thermo Scientific Chemicals

1-Chloro-3-fluoro-2-propanol, 95%

CAS: 453-11-2 | C3H6ClFO | 112.528 g/mol
Catalog NumberQuantity
L17286.06
also known as L17286-06
5 g
Catalog number L17286.06
also known as L17286-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS453-11-2
IUPAC Name1-chloro-3-fluoropropan-2-ol
Molecular FormulaC3H6ClFO
InChI KeyZNKJYZHZWALQNX-UHFFFAOYNA-N
SMILESOC(CF)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.4270-1.4330 @ 20?C
It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only