1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%

CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol
Quantity:
5 g
25 g
100 g
Catalog number L19086.14
also known as L19086-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS174501-64-5
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC8H15F6N2P
InChI KeyIXQYBUDWDLYNMA-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (unspecified)≥98.0%
Refractive Index1.4095-1.4135 @ 20?C
FormViscous liquid
Identification (FTIR)Conforms
View more
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.

Solubility
Miscible with dichloromethane, chloroform, ethyl acetate. Immiscible with water, diethyl ether and hexane.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Soni, S. K.; Sarkar, S.; Mirzadeh , N.; Selvakannan , P. R.; Bhargava, S. K. Self-Assembled Functional Nanostructure of Plasmid DNA with Ionic Liquid [Bmim][PF6]: Enhanced Efficiency in Bacterial Gene Transformation. Langmuir. 2015, 31 (16), 4722-4732.
  2. Zhang, Y.; Zhang, T.; Gan, P.; Li, H.; Zhang, M.; Jin, K.; Tang, S. Solubility of Isobutane in Ionic Liquids [BMIm][PF6], [BMIm][BF4], and [BMIm][Tf2N]. J. Chem. Eng. Data. 2015, 60 (6), 1706-1714.