4-Methoxy-3-pyrrolin-2-one, 99%
4-Methoxy-3-pyrrolin-2-one, 99%
4-Methoxy-3-pyrrolin-2-one, 99%
Thermo Scientific Chemicals

4-Methoxy-3-pyrrolin-2-one, 99%

CAS: 69778-83-2 | C5H7NO2 | 113.116 g/mol
Quantity:
1 g
5 g
Catalog number L19344.03
also known as L19344-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS69778-83-2
IUPAC Name4-methoxy-2,5-dihydro-1H-pyrrol-2-one
Molecular FormulaC5H7NO2
InChI KeyTXKQBYYDTLOLHA-UHFFFAOYSA-N
SMILESCOC1=CC(=O)NC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Melting Point (clear melt)128.0-135.0?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only