2,3-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
2,3-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
2,3-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Quantity:
250 mg
1 g
Catalog number L19445.03
also known as L19445-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein1622028
CAS1644-68-4
Chemical Name or Material2,3-Bis(trifluoromethyl)pyridine
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Flash Point70°C (158°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only