Petroleum Ether 40-60C CertiFied for analysis , Fisher Chemical™
Petroleum Ether 40-60C CertiFied for analysis , Fisher Chemical™
Petroleum Ether 40-60C CertiFied for analysis , Fisher Chemical™
Fisher Chemical™

Petroleum Ether 40-60C CertiFied for analysis , Fisher Chemical™

Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2%
Catalog number P-1760-27
also known as P/1760/27
Price (JPY)
-

Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2%

  • Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2% Plastic Bottle Cas No. 64742-49-0
Specifications
Boiling Point40.0°C to 65.0°C
CAS64742-49-0
Chemical Name or MaterialPetroleum
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
Melting Point<-100°C
PackagingPlastic
Quantity200 L
Unit SizeEach