Sea Sand, Extra Pure, SLR, Fisher Chemical™
Sea Sand, Extra Pure, SLR, Fisher Chemical™
Sea Sand, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

Sea Sand, Extra Pure, SLR, Fisher Chemical™

O2Si, CAS Number-14808-60-7, aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite, 5kg, 2230 deg.C, CHEBI:30563, Beige, 60.08g/mol, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 1610 deg.C, 11232, 60.083
Catalog NumberQuantity
S-0330-65
also known as S/0330/65
5 kg
Catalog number S-0330-65
also known as S/0330/65
Price (JPY)
-
Quantity:
5 kg
Request bulk or custom format
Specifications
Boiling Point2230°C
CAS14808-60-7
ChEBICHEBI:30563
ColorBeige
Formula Weight60.08g/mol
View more
O2Si, CAS Number-14808-60-7, aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite, 5kg, 2230 deg.C, CHEBI:30563, Beige, 60.08g/mol, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 1610 deg.C, 11232, 60.083