Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™
Fisher Chemical™

Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™

Catalog NumberQuantity
S-0365-60
also known as S/0365/60
1 kg
Catalog number S-0365-60
also known as S/0365/60
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Boiling Point2230°C
CAS14808-60-7
ChEBICHEBI:30563
ColorBeige
Formula Weight60.08g/mol
View more
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh