Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Fisher Chemical™

Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™

Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Powder Jar
Catalog number S-0761-60
also known as S/0761/60
Price (JPY)
-
Quantity:
1 kg
Packaging:
Plastic Powder Jar
Request bulk or custom format
Specifications
CAS7631-86-9
ChEBICHEBI:30563
ColorBrown-Orange
Formula Weight60.08g/mol
IUPAC Namesilanedione
View more
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent