Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

C2H9NaO5, CAS Number-6131-90-4, unii-4550k0sc9b, thomaegelin, plasmafusin, sodium acetate tn, sodium acetate usan:jan, tutofusin, sodium acetate usp:jan, natrium acetate-3-wasser, acetic acid, sodium salt, trihydrate, sodium acetate trihydrate, 5kg, CHEBI:32138, White, 136.08g/mol
Catalog NumberQuantityPackaging
S-2000-65
also known as S/2000/65
5 kgPlastic keg
Catalog number S-2000-65
also known as S/2000/65
Price (JPY)
-
Quantity:
5 kg
Packaging:
Plastic keg
Request bulk or custom format
Specifications
CAS6131-90-4
ChEBICHEBI:32138
ColorWhite
Formula Weight136.08g/mol
IUPAC Namesodium;acetate;trihydrate
View more
C2H9NaO5, CAS Number-6131-90-4, unii-4550k0sc9b, thomaegelin, plasmafusin, sodium acetate tn, sodium acetate usan:jan, tutofusin, sodium acetate usp:jan, natrium acetate-3-wasser, acetic acid, sodium salt, trihydrate, sodium acetate trihydrate, 5kg, CHEBI:32138, White, 136.08g/mol