Sodium chloride, meets analytical specification of Ph.Eur, Fisher Chemical™
Sodium chloride, meets analytical specification of Ph.Eur, Fisher Chemical™
Sodium chloride, meets analytical specification of Ph.Eur, Fisher Chemical™
Fisher Chemical™

Sodium chloride, meets analytical specification of Ph.Eur, Fisher Chemical™

ClNa, CAS Number-7647-14-5, salt, sodium chloride nacl, halite, common salt, table salt, sodium chloride, dendritis, saline, purex, rock salt, 5kg, 1461 deg.C, CHEBI:26710, White, 58.44g/mol, FAPWRFPIFSIZLT-UHFFFAOYSA-M, sodium;chloride, 801 deg.C, 3477, Plastic powder jar, 5.0 to 8.0, Solid, 5234 | CAS: 7647-14-5 | ClNa | 58.44 g/mol
Catalog NumberQuantityPackaging
S-3160-65
also known as S/3160/65
5 kgPlastic powder jar
Catalog number S-3160-65
also known as S/3160/65
Price (JPY)
-
Quantity:
5 kg
Packaging:
Plastic powder jar
ClNa, CAS Number-7647-14-5, salt, sodium chloride nacl, halite, common salt, table salt, sodium chloride, dendritis, saline, purex, rock salt, 5kg, 1461 deg.C, CHEBI:26710, White, 58.44g/mol, FAPWRFPIFSIZLT-UHFFFAOYSA-M, sodium;chloride, 801 deg.C, 3477, Plastic powder jar, 5.0 to 8.0, Solid, 5234
Specifications
Boiling Point1461°C
CAS7647-14-5
ChEBICHEBI:26710
ColorWhite
Formula Weight58.44g/mol
IUPAC Namesodium chloride
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
MDL NumberMFCD00003477
Melting Point801°C
Molecular FormulaClNa
PackagingPlastic powder jar
PubChem CID5234
SMILES[Na+].[Cl-]
Synonymsodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
Vapor Pressure1mmHg 865
Molecular Weight (g/mol)58.44
Physical FormSolid
Quantity5 kg
pH5.0 to 8.0
Unit SizeEach