Sodium chloride, Fisher Chemical™
Sodium chloride, Fisher Chemical™
Fisher Chemical™

Sodium chloride, Fisher Chemical™

Sodium chloride Certified AR ACS EP
Catalog number S-3160-70
also known as S/3160/70
Price (JPY)
-
Contact Us ›
Sodium chloride Certified AR ACS EP
  • Sodium chloride
  • Certified AR
  • ACS EP
  • Analytical reagent grade
  • Plastic Bottle
  • CAS No. 7647-14-5
Specifications
Boiling Point100°C
CAS7647-14-5
Chemical Name or MaterialSodium chloride
IUPAC Namesodium chloride
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
MDL NumberMFCD00003477
Melting Point801°C (literature)
Molecular FormulaClNa
PackagingPlastic Bottle
SMILES[Na+].[Cl-]
Molecular Weight (g/mol)58.44
Quantity25 kg
Unit SizeEach