Tri-Sodium Citrate Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Tri-Sodium Citrate Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Tri-Sodium Citrate Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Fisher Chemical™

Tri-Sodium Citrate Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Catalog NumberQuantityPackaging
S-3320-53
also known as S/3320/53
500 gPlastic powder jar
Catalog number S-3320-53
also known as S/3320/53
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS4-3-6132
ChEBICHEBI:32142
ColorWhite
Formula Weight294.09g/mol
IUPAC Nametrisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
View more
Tri-Sodium Citrate Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP