Sodium Fluoride, 99.0+%, Certified AR for Analysis, Fisher Chemical™
Sodium Fluoride, 99.0+%, Certified AR for Analysis, Fisher Chemical™
Sodium Fluoride, 99.0+%, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Sodium Fluoride, 99.0+%, Certified AR for Analysis, Fisher Chemical™

Catalog NumberQuantity
S-3920-50
also known as S/3920/50
250 g
Catalog number S-3920-50
also known as S/3920/50
Price (JPY)
-
Quantity:
250 g
Request bulk or custom format
Specifications
Boiling Point1700°C
CAS7681-49-4
ChEBICHEBI:28741
ColorWhite
Formula Weight41.98g/mol
View more

Sodium Fluoride, 99.0+%, Certified AR for Analysis