Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Fisher Chemical™

Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M
Catalog NumberQuantityPackaging
S-4240-53
also known as S/4240/53
500 gPlastic powder jar
Catalog number S-4240-53
also known as S/4240/53
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS144-55-8
ChEBICHEBI:32139
ColorWhite
Formula Weight84.01g/mol
IUPAC Namesodium;hydrogen carbonate
View more
CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M