Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™

Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, Packaging: Plastic powder jar, CAS: 10028-24-7, White, Melting Point: 243 deg.C, Molecular Formula: H5Na2O6P, Molecular Weight: 177.987, pH: 8.7 to 9.4
Catalog NumberQuantityPackaging
S-4450-53
also known as S/4450/53
500 gPlastic powder jar
Catalog number S-4450-53
also known as S/4450/53
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS10028-24-7
ColorWhite
Formula Weight177.99g/mol
IUPAC Namedisodium;hydrogen phosphate;dihydrate
InChI KeyKDQPSPMLNJTZAL-UHFFFAOYSA-L
View more
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, Packaging: Plastic powder jar, CAS: 10028-24-7, White, Melting Point: 243 deg.C, Molecular Formula: H5Na2O6P, Molecular Weight: 177.987, pH: 8.7 to 9.4