Di-Sodium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Di-Sodium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Di-Sodium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

HNa2O4P, CAS Number-7558-79-4, disodium orthophosphate, acetest, disodium hydrogen phosphate, disodium hydrogenorthophosphate, disodium phosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt, soda phosphate, sodium phosphate dibasic, dibasic sodium phosphate, 3kg, CHEBI:34683, White
Quantity:
500 g
1 kg
3 kg
Packaging:
Plastic powder jar
Catalog number S-4520-60
also known as S/4520/60
Price (JPY)
-
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS7558-79-4
ChEBICHEBI:34683
ColorWhite
Formula Weight141.96g/mol
IUPAC Namedisodium hydrogen phosphate
View more
HNa2O4P, CAS Number-7558-79-4, disodium orthophosphate, acetest, disodium hydrogen phosphate, disodium hydrogenorthophosphate, disodium phosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt, soda phosphate, sodium phosphate dibasic, dibasic sodium phosphate, 3kg, CHEBI:34683, White