Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548 | CAS: 1310-73-2 | HNaO | 39.997 g/mol
Quantity:
500 g
1 kg
3 kg
5 kg
Catalog number S-4920-53
also known as S/4920/53
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point1390°C
CAS1310-73-2
ChEBICHEBI:32145
ColorWhite
Formula Weight40g/mol
View more
HNaO, CAS Number-1310-73-2, white caustic, aetznatron, sodium hydroxide na oh, caustic soda, ascarite, soda lye, soda, caustic, sodium hydroxide solution, sodium hydrate, sodium hydroxide, 5kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548