Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™
Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™

Na2O4S, CAS Number-7757-82-6, disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite, 5kg, CHEBI:32149, White, 142.04g/mol, PMZURENOXWZQFD-UHFFFAOYSA-L
Quantity:
500 g
5 kg
Catalog number S-6640-65
also known as S/6640/65
Price (JPY)
-
Quantity:
5 kg
Request bulk or custom format
Specifications
CAS7757-82-6
ChEBICHEBI:32149
ColorWhite
Formula Weight142.04g/mol
IUPAC Namedisodium sulfate
View more
Na2O4S, CAS Number-7757-82-6, disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite, 5kg, CHEBI:32149, White, 142.04g/mol, PMZURENOXWZQFD-UHFFFAOYSA-L