Chemical Binder Spillage Treatment, Pure, Fisher Chemical™
Chemical Binder Spillage Treatment, Pure, Fisher Chemical™
Fisher Chemical™
Chemical Binder Spillage Treatment, Pure, Fisher Chemical™
Universally applicable chemical binder suitable for use with all chemical spills.
Catalog NumberPackaging Quantity
S-7435-53
also known as S/7435/53
500 g
Catalog number S-7435-53
also known as S/7435/53
Price (JPY)
-
Contact Us ›
Packaging Quantity:
500 g
Universally applicable chemical binder suitable for use with all chemical spills.
  • Binds with liquids with a volume up to 75 times its own weight
  • Does not eliminate the hazards associated with a spillage and the residue must be handled and disposed of according to the substance bound
Specifications
ColorGreen
Packaging Quantity500 g
Unit SizeEach