Starch, Soluble, Technical, SLR, Fisher Chemical™
Starch, Soluble, Technical, SLR, Fisher Chemical™
Starch, Soluble, Technical, SLR, Fisher Chemical™
Fisher Chemical™

Starch, Soluble, Technical, SLR, Fisher Chemical™

Catalog NumberQuantity
S-7960-50
also known as S/7960/50
250 g
Catalog number S-7960-50
also known as S/7960/50
Price (JPY)
-
Quantity:
250 g
Request bulk or custom format
Specifications
CAS9005-25-8
ChEBICHEBI:18167
ColorWhite
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
InChI KeyGUBGYTABKSRVRQ-UHFFFAOYNA-N
View more
Starch, Soluble, Technical, SLR