Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

C12H22O11, CAS Number-57-50-1, amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar, 1kg, CHEBI:17992, White, 342.29g/mol, CZMRCDWAGMRECN-UGDNZRGBSA-N | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic powder jar
Catalog number S-8600-53
also known as S/8600/53
Price (JPY)
-
Contact Us ›
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS57-50-1
ChEBICHEBI:17992
ColorWhite
Formula Weight342.29g/mol
IUPAC Name(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
View more
C12H22O11, CAS Number-57-50-1, amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar, 1kg, CHEBI:17992, White, 342.29g/mol, CZMRCDWAGMRECN-UGDNZRGBSA-N