Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Fisher Chemical™

Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Quantity:
1 L
2.5 L
Packaging:
HDPE plastic bottle
Catalog number S-9231-PB15
also known as S/9231/PB15
Price (JPY)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Boiling Point290°C
CAS7664-93-9
ChEBICHEBI:26836
ColorColorless
Formula Weight98.07g/mol
View more
Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis