Sulfuric Acid Min 95% d=1.83, Certified AR, for Analysis, Fisher Chemical™
Sulfuric Acid Min 95% d=1.83, Certified AR, for Analysis, Fisher Chemical™
Sulfuric Acid Min 95% d=1.83, Certified AR, for Analysis, Fisher Chemical™
Fisher Chemical™

Sulfuric Acid Min 95% d=1.83, Certified AR, for Analysis, Fisher Chemical™

H2O4S, CAS Number-7664-93-9, electrolyte acid, acidum sulfuricum, battery acid, dihydrogen sulfate, sulphuric acid, oil of vitriol, acide sulfurique, mattling acid, vitriol brown oil, dipping acid, 2.5L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid | CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
HDPE plastic bottle
Catalog number S-9240-PC17
also known as S/9240/PC17
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
Boiling Point290°C
CAS7664-93-9
ChEBICHEBI:26836
ColorColorless
Formula Weight98.07g/mol
View more
H2O4S, CAS Number-7664-93-9, electrolyte acid, acidum sulfuricum, battery acid, dihydrogen sulfate, sulphuric acid, oil of vitriol, acide sulfurique, mattling acid, vitriol brown oil, dipping acid, 2.5L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid