Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Fisher Chemical™

Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid

O12S3Ti2, CAS Number-10343-61-0, 69hci1s2xi, acmc-1bolh, dititanium tris sulphate, sulfuric acid, titanium 3+ salt 3:2, titan iii-sulfat, titanium 3+ sulfate 2:3, titanium iii sulfate, titanium sesquisulfate, unii-69hci1s2xi, 500mL, Purple, SOBXOQKKUVQETK-UHFFFAOYSA-H, titanium(3+);trisulfate, 36317
Catalog NumberQuantity
T-2000-PB08
also known as T/2000/PB08
500 mL
Catalog number T-2000-PB08
also known as T/2000/PB08
Price (JPY)
-
Quantity:
500 mL
O12S3Ti2, CAS Number-10343-61-0, 69hci1s2xi, acmc-1bolh, dititanium tris sulphate, sulfuric acid, titanium 3+ salt 3:2, titan iii-sulfat, titanium 3+ sulfate 2:3, titanium iii sulfate, titanium sesquisulfate, unii-69hci1s2xi, 500mL, Purple, SOBXOQKKUVQETK-UHFFFAOYSA-H, titanium(3+);trisulfate, 36317
Specifications
CAS10343-61-0
ColorPurple
IUPAC Nametitanium(3+);trisulfate
InChI KeySOBXOQKKUVQETK-UHFFFAOYSA-H
MDL Number36317
Molecular FormulaO12S3Ti2
PackagingPlastic Coated Glass bottle
Physical FormLiquid
PubChem CID165866
SMILES[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ti+3].[Ti+3]
Synonymtitanium sesquisulfate, titanium iii sulfate, unii-69hci1s2xi, 69hci1s2xi, dititanium tris sulphate, titanium 3+ sulfate 2:3, titan iii-sulfat, acmc-1bolh, sulfuric acid, titanium 3+ salt 3:2
Molecular Weight (g/mol)383.902
Quantity500 mL
pH1
Unit SizeEach