Toluene, 99+%, Extra Pure, SLR, Fisher Chemical™
Toluene, 99+%, Extra Pure, SLR, Fisher Chemical™
Toluene, 99+%, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

Toluene, 99+%, Extra Pure, SLR, Fisher Chemical™

Catalog NumberQuantityPackaging
T-2200-08
also known as T/2200/08
500 mLAmber Glass Bottle
Catalog number T-2200-08
also known as T/2200/08
Price (JPY)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Boiling Point111°C
CAS108-88-3
ChEBICHEBI:17578
ColorColorless
Formula Weight92.14g/mol
View more

Toluene, 99+%, Extra Pure, SLR