Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™
Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™
Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™
Fisher Chemical™

Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™

Quantity:
100 mL
500 mL
Catalog number T-3255-PB05
also known as T/3255/PB05
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Boiling Point72°C
CAS76-05-1
ChEBICHEBI:45892
ColorColorless
Formula Weight114.02g/mol
View more
Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing