Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™
Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™
Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™

Quantity:
100 g
1 kg
Catalog number T-3710-60
also known as T/3710/60
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Boiling Point219°C
CAS77-86-1
ChEBICHEBI:9754
ColorWhite
Formula Weight121.14g/mol
View more

Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR