Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03306842_pri
          Assay Name:
          mmu-mir-24-2
          Stem-loop ID:
          mmu-mir-24-2 mmu-mir-3074-2
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.8: 84208815 - 84208921 [+] on Build GRCm38
          Stem-loop Location:
          Chr.8: 84208815 - 84208921 [+] on Build GRCm38 Chr.8: 84208825 - 84208907 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-24-3p mmu-miR-24-2-5p mmu-miR-3074-5p mmu-miR-3074-2-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03306842_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          71

          Chromosome Location:

          Chr. 8 - 84209116 - 84209116 [+] on 84209116 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-24-3p MIMAT0000219
          Mouse mmu-miR-24-2-5p MIMAT0005440
          Mouse mmu-miR-3074-5p MIMAT0014856
          Mouse mmu-miR-3074-2-3p MIMAT0014946

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-24-2
          Stem-loop Accession # MI0000572
          Stem-loop Sequence
          GCCUCUCUCCGGGCUCCGCCUCCCGUGCCUACUGAGCUGAAACAGUUGAUUCCAGUGCACUGGCUCAGUUCAGCAGGAACAGGAGUCCAGCCCCCUAGGAGCUGGCA
          Chromosome Location Chr. 8 - 84208815 - 84208921 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000219
          miRBase ID: mmu-miR-24-3p
          Mature miRNA Sequence: UGGCUCAGUUCAGCAGGAACAG
          Chromosome Location: Chr. 8 - 84208815 - 84208921 [+] on Build GRCm38

          miRBase Accession #: MIMAT0005440
          miRBase ID: mmu-miR-24-2-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAAACAGU
          Chromosome Location: Chr. 8 - 84208815 - 84208921 [+] on Build GRCm38

          Gene Family ID MIPF0000041, mir-24
          Distance to Amplicon 157 bases
          Stem-loop ID mmu-mir-3074-2
          Stem-loop Accession # MI0014104
          Stem-loop Sequence
          GGGCUGGACUCCUGUUCCUGCUGAACUGAGCCAGUGCACUGGAAUCAACUGUUUCAGCUCAGUAGGCACGGGAGGCGGAGCCC
          Chromosome Location Chr. 8 - 84208825 - 84208907 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014856
          miRBase ID: mmu-miR-3074-5p
          Mature miRNA Sequence: GUUCCUGCUGAACUGAGCCAGU
          Chromosome Location: Chr. 8 - 84208825 - 84208907 [-] on Build GRCm38

          miRBase Accession #: MIMAT0014946
          miRBase ID: mmu-miR-3074-2-3p
          Mature miRNA Sequence: UGUUUCAGCUCAGUAGGCAC
          Chromosome Location: Chr. 8 - 84208825 - 84208907 [-] on Build GRCm38

          Gene Family ID MIPF0001103, mir-3074
          Distance to Amplicon 171 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10737
          Ambion® Pre-miR™ miRNA Precursor : PM10737
          TaqMan™ MicroRNA Assay : 000402
          mirVana® miRNA inhibitor : MH10737
          mirVana® miRNA mimic : MC10737
          TaqMan™ Advanced miRNA Assay : 477992_mir
          TaqMan™ Advanced miRNA Assay : rno481011_mir
          TaqMan™ Advanced miRNA Assay : mmu481011_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20237
          Ambion® Pre-miR™ miRNA Precursor : PM20237
          TaqMan™ MicroRNA Assay : 462202_mat
          mirVana® miRNA inhibitor : MH20237
          mirVana® miRNA mimic : MC20237
          TaqMan™ Advanced miRNA Assay : mmu481705_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19968
          Ambion® Pre-miR™ miRNA Precursor : PM19968
          TaqMan™ MicroRNA Assay : 464324_mat
          mirVana® miRNA inhibitor : MH19968
          mirVana® miRNA mimic : MC19968
          TaqMan™ Advanced miRNA Assay : mmu482640_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12270
          Ambion® Pre-miR™ miRNA Precursor : PM12270
          TaqMan™ MicroRNA Assay : 002494
          mirVana® miRNA inhibitor : MH12270
          mirVana® miRNA mimic : MC12270
          TaqMan™ Advanced miRNA Assay : mmu482591_mir
          TaqMan™ Advanced miRNA Assay : rno481447_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline