Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno479228_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-19a-3p
          Stem-loop Accession Number: MI0000073
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 91350891 - 91350972 [+] on Build GRCh38
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Species: Human, Mouse, Rat, Ateles geoffroyi, Bovine, Callithrix jacchus, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Goat, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Lemur catta, Macaca nemestrina, Monodelphis domestica, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Saguinus labiatus, Tetraodon nigroviridis, Tupaia chinensis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno479228_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000011, mir-19

          Mature miRNA Sequence:

          UGUGCAAAUCUAUGCAAAACUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-19a-3p MIMAT0000073
          Mouse mmu-miR-19a-3p MIMAT0000651
          Rat rno-miR-19a-3p MIMAT0000789
          Ateles geoffroyi age-miR-19a MIMAT0002675
          Bovine bta-miR-19a MIMAT0004336
          Callithrix jacchus cja-miR-19a MIMAT0049329
          View More Rows
          Chicken gga-miR-19a-3p MIMAT0001112
          Columba livia cli-miR-19a-3p MIMAT0038428
          Cricetulus griseus cgr-miR-19a MIMAT0023839
          Dasypus novemcinctus dno-miR-19a-3p MIMAT0047516
          Dog cfa-miR-19a MIMAT0006650
          Fugu rubripes fru-miR-19a MIMAT0002920
          Gadus morhua gmo-miR-19a-3p MIMAT0044303
          Goat chi-miR-19a MIMAT0036042
          Gorilla gorilla ggo-miR-19a MIMAT0002661
          Guinea pig cpo-miR-19a-3p MIMAT0046864
          Horse eca-miR-19a MIMAT0013086
          Lagothrix lagotricha lla-miR-19a MIMAT0002717
          Lemur catta lca-miR-19a MIMAT0002668
          Macaca nemestrina mne-miR-19a MIMAT0002724
          Monodelphis domestica mdo-miR-19a-3p MIMAT0004168
          Nomascus leucogenys nle-miR-19a MIMAT0049454
          Ophiophagus hannah oha-miR-19a-3p MIMAT0036796
          Ornithorhynchus anatinus oan-miR-19a-3p MIMAT0007148
          Pan paniscus ppa-miR-19a MIMAT0002682
          Pan troglodytes ptr-miR-19a MIMAT0002696
          Pig ssc-miR-19a MIMAT0002128
          Pongo pygmaeus ppy-miR-19a MIMAT0002689
          Python bivittatus pbv-miR-19a-3p MIMAT0038844
          Rabbit ocu-miR-19a-3p MIMAT0048116
          Rhesus monkey mml-miR-19a-3p MIMAT0002703
          Saguinus labiatus sla-miR-19a MIMAT0002710
          Tetraodon nigroviridis tni-miR-19a MIMAT0002921
          Tupaia chinensis tch-miR-19a-3p MIMAT0036603
          Xenopus tropicalis xtr-miR-19a MIMAT0003566
          Zebrafish dre-miR-19a-3p MIMAT0001782

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-19a
          Stem-loop Accession # MI0000073
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location Chr. 13 - 91350891 - 91350972 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000073
          miRBase ID: hsa-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: Chr. 13 - 91350891 - 91350972 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-19a
          Stem-loop Accession # MI0000688
          Stem-loop Sequence
          GCAGCCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location Chr. 14 - 115044000 - 115044081 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000651
          miRBase ID: mmu-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: Chr. 14 - 115044000 - 115044081 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-19a
          Stem-loop Accession # MI0000849
          Stem-loop Sequence
          GCAGCCCUCUGUUCGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location Chr. 15 - 100180162 - 100180243 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000789
          miRBase ID: rno-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: Chr. 15 - 100180162 - 100180243 [+] on Build Rnor_6.0

          Ateles geoffroyi

          Stem-loop ID age-mir-19a
          Stem-loop Accession # MI0002979
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGCAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002675
          miRBase ID: age-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-19a
          Stem-loop Accession # MI0005460
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004336
          miRBase ID: bta-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-19a
          Stem-loop Accession # MI0040065
          Stem-loop Sequence
          AGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049329
          miRBase ID: cja-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-19a
          Stem-loop Accession # MI0001182
          Stem-loop Sequence
          GCAGUCUUCUGUUAGUUUUGCAUAGUUGCACUACAGGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001112
          miRBase ID: gga-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-19a
          Stem-loop Accession # MI0029915
          Stem-loop Sequence
          UUUGCAGUCUUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAGUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038428
          miRBase ID: cli-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-19a
          Stem-loop Accession # MI0020450
          Stem-loop Sequence
          GCAGCCCUCUGUUAGUUUUGCAUACUUGCACUACAAGAAGAAUGCAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023839
          miRBase ID: cgr-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-19a
          Stem-loop Accession # MI0038912
          Stem-loop Sequence
          AGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047516
          miRBase ID: dno-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-19a
          Stem-loop Accession # MI0008051
          Stem-loop Sequence
          AGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006650
          miRBase ID: cfa-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-19a
          Stem-loop Accession # MI0003235
          Stem-loop Sequence
          GCAGUUCUCUGCUAGUUUUGCAUAGUUGCACUACAAGAAUAGAGGAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002920
          miRBase ID: fru-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-19a-1
          Stem-loop Accession # MI0036195
          Stem-loop Sequence
          AGUUCUGCAUAGUUGCACUACAAGAAUAGAAGAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044303
          miRBase ID: gmo-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Stem-loop ID gmo-mir-19a-2
          Stem-loop Accession # MI0036217
          Stem-loop Sequence
          AGUUUUGCGUAGUUGCUCUGCAAGAAAAUUGAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044303
          miRBase ID: gmo-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-19a
          Stem-loop Accession # MI0030677
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036042
          miRBase ID: chi-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-19a
          Stem-loop Accession # MI0002967
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002661
          miRBase ID: ggo-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-19a
          Stem-loop Accession # MI0038568
          Stem-loop Sequence
          AGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046864
          miRBase ID: cpo-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-19a
          Stem-loop Accession # MI0012833
          Stem-loop Sequence
          GCAGUCUUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013086
          miRBase ID: eca-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-19a
          Stem-loop Accession # MI0003015
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGCAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002717
          miRBase ID: lla-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Lemur catta

          Stem-loop ID lca-mir-19a
          Stem-loop Accession # MI0002973
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002668
          miRBase ID: lca-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-19a
          Stem-loop Accession # MI0003021
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAGGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002724
          miRBase ID: mne-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-19a
          Stem-loop Accession # MI0005356
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004168
          miRBase ID: mdo-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-19a
          Stem-loop Accession # MI0040206
          Stem-loop Sequence
          AGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049454
          miRBase ID: nle-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-19a
          Stem-loop Accession # MI0031395
          Stem-loop Sequence
          UUUUGCAGUCUUCUGUUAGUUUUGCAUAGUUGCACUACAGGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUUGUGGCCUGUUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036796
          miRBase ID: oha-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-19a
          Stem-loop Accession # MI0006873
          Stem-loop Sequence
          UUGGGGUGGUCUUGUUUUUGUUUGUUUUUUUUUUUUUUUGUUUGCAGCUCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGACGGUGGCCUGCUAUUUCCUACAUCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007148
          miRBase ID: oan-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-19a
          Stem-loop Accession # MI0002985
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002682
          miRBase ID: ppa-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-19a
          Stem-loop Accession # MI0002997
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002696
          miRBase ID: ptr-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-19a
          Stem-loop Accession # MI0002422
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002128
          miRBase ID: ssc-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-19a
          Stem-loop Accession # MI0002991
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002689
          miRBase ID: ppy-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-19a
          Stem-loop Accession # MI0030164
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAGGAAGAGUGUAGUUGUGCAAAUCUAUGCAAAACUGAUUGUGGCCUGUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038844
          miRBase ID: pbv-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-19a
          Stem-loop Accession # MI0039228
          Stem-loop Sequence
          AGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048116
          miRBase ID: ocu-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-19a
          Stem-loop Accession # MI0003003
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002703
          miRBase ID: mml-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-19a
          Stem-loop Accession # MI0003009
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002710
          miRBase ID: sla-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-19a
          Stem-loop Accession # MI0003236
          Stem-loop Sequence
          GCAGUUCUCUGCUAGUUUUGCAUAGUUGCACUACAAGAAUAGAGGAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002921
          miRBase ID: tni-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-19a
          Stem-loop Accession # MI0031271
          Stem-loop Sequence
          UAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036603
          miRBase ID: tch-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-19a
          Stem-loop Accession # MI0004804
          Stem-loop Sequence
          GCAGUCUUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAAAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGCAGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003566
          miRBase ID: xtr-miR-19a
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-19a
          Stem-loop Accession # MI0001903
          Stem-loop Sequence
          UGGUGCAGUUCUCUGCUAGUUUUGCAUAGUUGCACUACAAGAAAACGGGAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001782
          miRBase ID: dre-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04228426_pri
          TaqMan™ Pri-miRNA Assay : Hs03295901_pri
          TaqMan™ Pri-miRNA Assay : Rn03465733_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10649
          Ambion® Pre-miR™ miRNA Precursor : PM10649
          TaqMan™ MicroRNA Assay : 000395
          mirVana® miRNA inhibitor : MH10649
          mirVana® miRNA mimic : MC10649
          TaqMan™ Advanced miRNA Assay : mmu479228_mir
          TaqMan™ Advanced miRNA Assay : 479228_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline