Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03295901_pri
          Assay Name:
          hsa-mir-17
          Stem-loop ID:
          hsa-mir-17 hsa-mir-18a hsa-mir-19a
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.13: 91350605 - 91350688 [+] on Build GRCh38
          Stem-loop Location:
          Chr.13: 91350605 - 91350688 [+] on Build GRCh38 Chr.13: 91350751 - 91350821 [+] on Build GRCh38 Chr.13: 91350891 - 91350972 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-17-5p hsa-miR-17-3p hsa-miR-18a-3p hsa-miR-18a-5p hsa-miR-19a-5p hsa-miR-19a-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03295901_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          100

          Chromosome Location:

          Chr. 13 - 91350441 - 91350441 [+] on 91350441 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-17-5p MIMAT0000070
          Human hsa-miR-17-3p MIMAT0000071
          Human hsa-miR-18a-3p MIMAT0002891
          Human hsa-miR-18a-5p MIMAT0000072
          Human hsa-miR-19a-5p MIMAT0004490
          Human hsa-miR-19a-3p MIMAT0000073

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-17
          Stem-loop Accession # MI0000071
          Stem-loop Sequence
          GUCAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGCAUUAUGGUGAC
          Chromosome Location Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000070
          miRBase ID: hsa-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38

          miRBase Accession #: MIMAT0000071
          miRBase ID: hsa-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 103 bases
          Stem-loop ID hsa-mir-18a
          Stem-loop Accession # MI0000072
          Stem-loop Sequence
          UGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCA
          Chromosome Location Chr. 13 - 91350751 - 91350821 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002891
          miRBase ID: hsa-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUGG
          Chromosome Location: Chr. 13 - 91350751 - 91350821 [+] on Build GRCh38

          miRBase Accession #: MIMAT0000072
          miRBase ID: hsa-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: Chr. 13 - 91350751 - 91350821 [+] on Build GRCh38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 249 bases
          Stem-loop ID hsa-mir-19a
          Stem-loop Accession # MI0000073
          Stem-loop Sequence
          GCAGUCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location Chr. 13 - 91350891 - 91350972 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004490
          miRBase ID: hsa-miR-19a-5p
          Mature miRNA Sequence: AGUUUUGCAUAGUUGCACUACA
          Chromosome Location: Chr. 13 - 91350891 - 91350972 [+] on Build GRCh38

          miRBase Accession #: MIMAT0000073
          miRBase ID: hsa-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: Chr. 13 - 91350891 - 91350972 [+] on Build GRCh38

          Gene Family ID MIPF0000011, mir-19
          Distance to Amplicon 389 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12412
          Ambion® Pre-miR™ miRNA Precursor : PM12412
          TaqMan™ MicroRNA Assay : 002308
          mirVana® miRNA inhibitor : MH12412
          mirVana® miRNA mimic : MC12412
          TaqMan™ Advanced miRNA Assay : 478447_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12264
          Ambion® Pre-miR™ miRNA Precursor : PM12264
          TaqMan™ MicroRNA Assay : 002423
          mirVana® miRNA inhibitor : MH12264
          mirVana® miRNA mimic : MC12264
          TaqMan™ Advanced miRNA Assay : 477944_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12420
          Ambion® Pre-miR™ miRNA Precursor : PM12420
          TaqMan™ MicroRNA Assay : 002424
          mirVana® miRNA inhibitor : MH12420
          mirVana® miRNA mimic : MC12420
          TaqMan™ Advanced miRNA Assay : 478750_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12246
          Ambion® Pre-miR™ miRNA Precursor : PM12246
          TaqMan™ MicroRNA Assay : 002421
          mirVana® miRNA inhibitor : MH12246
          mirVana® miRNA mimic : MC12246
          TaqMan™ Advanced miRNA Assay : 477932_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12973
          Ambion® Pre-miR™ miRNA Precursor : PM12973
          TaqMan™ MicroRNA Assay : 002422
          mirVana® miRNA inhibitor : MH12973
          mirVana® miRNA mimic : MC12973
          TaqMan™ Advanced miRNA Assay : 478551_mir
          TaqMan™ Advanced miRNA Assay : rno480968_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10649
          Ambion® Pre-miR™ miRNA Precursor : PM10649
          TaqMan™ MicroRNA Assay : 000395
          mirVana® miRNA inhibitor : MH10649
          mirVana® miRNA mimic : MC10649
          TaqMan™ Advanced miRNA Assay : mmu479228_mir
          TaqMan™ Advanced miRNA Assay : rno479228_mir
          TaqMan™ Advanced miRNA Assay : 479228_mir


          miRBase Alias:

          Human: hsa-mir-18a (v21)
          Human: hsa-mir-19a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline