Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12973
          Assay Name: hsa-miR-18a-5p
          miRBase Accession Number: MI0000072
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 91350751 - 91350821 [+] on Build GRCh38
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Goat, Guinea pig, Horse, Microcebus murinus, Ophiophagus hannah, Oreochromis niloticus, Oryzias latipes, Otolemur garnettii, Petromyzon marinus, Python bivittatus, Rabbit, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12973

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          UAAGGUGCAUCUAGUGCAGAUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-18a-5p MIMAT0000072
          Mouse mmu-miR-18a-5p MIMAT0000528
          Rat rno-miR-18a-5p MIMAT0000787
          Alligator mississippiensis ami-miR-18-5p MIMAT0038049
          Anolis carolinensis aca-miR-18a-5p MIMAT0021817
          Cricetulus griseus cgr-miR-18a-5p MIMAT0023817
          View More Rows
          Cyprinus carpio ccr-miR-18a MIMAT0026246
          Dasypus novemcinctus dno-miR-18a-5p MIMAT0047511
          Goat chi-miR-18a-5p MIMAT0036015
          Guinea pig cpo-miR-18a-5p MIMAT0046859
          Horse eca-miR-18a MIMAT0013085
          Microcebus murinus mmr-miR-18 MIMAT0049197
          Ophiophagus hannah oha-miR-18a-5p MIMAT0036768
          Oreochromis niloticus oni-miR-18a-2 MIMAT0043044
          Oryzias latipes ola-miR-18 MIMAT0022532
          Otolemur garnettii oga-miR-18 MIMAT0049647
          Petromyzon marinus pma-miR-18a-5p MIMAT0019387
          Python bivittatus pbv-miR-18a-5p MIMAT0038840
          Rabbit ocu-miR-18a-5p MIMAT0048111
          Xenopus tropicalis xtr-miR-18a-5p MIMAT0003652

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-18a
          Stem-loop Accession # MI0000072
          Stem-loop Sequence
          UGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCA
          Chromosome Location Chr. 13 - 91350751 - 91350821 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000072
          miRBase ID: hsa-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: Chr. 13 - 91350751 - 91350821 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-18a
          Stem-loop Accession # MI0000567
          Stem-loop Sequence
          UGCGUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGACUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUGUC
          Chromosome Location Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000528
          miRBase ID: mmu-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-18a
          Stem-loop Accession # MI0000846
          Stem-loop Sequence
          UGCGUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGACUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUGUC
          Chromosome Location Chr. 15 - 100180015 - 100180110 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000787
          miRBase ID: rno-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: Chr. 15 - 100180015 - 100180110 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-18
          Stem-loop Accession # MI0029673
          Stem-loop Sequence
          GCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038049
          miRBase ID: ami-miR-18-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-18a
          Stem-loop Accession # MI0018783
          Stem-loop Sequence
          AGUGACUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021817
          miRBase ID: aca-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-18a
          Stem-loop Accession # MI0020433
          Stem-loop Sequence
          GUGCUGAGUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGACUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUGUCGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023817
          miRBase ID: cgr-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-18a
          Stem-loop Accession # MI0023350
          Stem-loop Sequence
          CUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGACUAGCACCUACUGCCCUAAGUGCUCCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026246
          miRBase ID: ccr-miR-18a
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-18a
          Stem-loop Accession # MI0038910
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047511
          miRBase ID: dno-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-18a
          Stem-loop Accession # MI0030660
          Stem-loop Sequence
          GCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036015
          miRBase ID: chi-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-18a
          Stem-loop Accession # MI0038566
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046859
          miRBase ID: cpo-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-18a
          Stem-loop Accession # MI0012832
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013085
          miRBase ID: eca-miR-18a
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-18
          Stem-loop Accession # MI0039917
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049197
          miRBase ID: mmr-miR-18
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-18a
          Stem-loop Accession # MI0031381
          Stem-loop Sequence
          UUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAAUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036768
          miRBase ID: oha-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-18a-2
          Stem-loop Accession # MI0035049
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAAUAGACUAGCACCUACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043044
          miRBase ID: oni-miR-18a-2
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-18-1
          Stem-loop Accession # MI0019417
          Stem-loop Sequence
          CAUACGAUGUACUGUCUUUGUGCUAAGGUGCAUCUAGUGCAGAUAGUGAAAUAGACUAGCACCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGGGGCUGUAAUAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022532
          miRBase ID: ola-miR-18
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Stem-loop ID ola-mir-18-2
          Stem-loop Accession # MI0019541
          Stem-loop Sequence
          UCUAGUGCUUAUGCUAAGGUGCAUCUAGUGCAGAUAGUGAAAUAGACUAGCACCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGGAGCUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022532
          miRBase ID: ola-miR-18
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-18
          Stem-loop Accession # MI0040430
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049647
          miRBase ID: oga-miR-18
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-18a
          Stem-loop Accession # MI0017017
          Stem-loop Sequence
          CCUGUGCUUGCUUGUGCUAAGGUGCAUCUAGUGCAGAUAGUUCCGCUGCGUAAAUGCUACUGCUCUAUGUGCUCCUUUACCCACAAGAGGUCACGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019387
          miRBase ID: pma-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-18a
          Stem-loop Accession # MI0030162
          Stem-loop Sequence
          AUAACUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038840
          miRBase ID: pbv-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-18a
          Stem-loop Accession # MI0039226
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048111
          miRBase ID: ocu-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-18a
          Stem-loop Accession # MI0004893
          Stem-loop Sequence
          GUGCUUUUUGUCCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAAAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003652
          miRBase ID: xtr-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04228426_pri
          TaqMan™ Pri-miRNA Assay : Hs03295901_pri
          TaqMan™ Pri-miRNA Assay : Rn03465733_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12973
          Ambion® Pre-miR™ miRNA Precursor : PM12973
          TaqMan™ MicroRNA Assay : 002422
          mirVana® miRNA mimic : MC12973
          TaqMan™ Advanced miRNA Assay : 478551_mir
          TaqMan™ Advanced miRNA Assay : rno480968_mir


          miRBase Alias:

          Human: hsa-miR-18a (v17)
          Mouse: mmu-miR-18a (v17)
          Rat: rno-miR-18a (v18)
          Aeromonas caviae Ae398: aca-miR-18a (v18)
          Phocea massiliensis: pma-miR-18a (v18)
          Xenopus tropicalis: xtr-miR-18a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline