Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04228426_pri
          Assay Name:
          mmu-mir-17
          Stem-loop ID:
          mmu-mir-17 mmu-mir-18a mmu-mir-19a
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.14: 115043671 - 115043754 [+] on Build GRCm38
          Stem-loop Location:
          Chr.14: 115043671 - 115043754 [+] on Build GRCm38 Chr.14: 115043851 - 115043946 [+] on Build GRCm38 Chr.14: 115044000 - 115044081 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-17-3p mmu-miR-17-5p mmu-miR-18a-5p mmu-miR-18a-3p mmu-miR-19a-5p mmu-miR-19a-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm04228426_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          118

          Chromosome Location:

          Chr. 14 - 115043501 - 115043501 [+] on 115043501 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-17-3p MIMAT0000650
          Mouse mmu-miR-17-5p MIMAT0000649
          Mouse mmu-miR-18a-5p MIMAT0000528
          Mouse mmu-miR-18a-3p MIMAT0004626
          Mouse mmu-miR-19a-5p MIMAT0004660
          Mouse mmu-miR-19a-3p MIMAT0000651

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-17
          Stem-loop Accession # MI0000687
          Stem-loop Sequence
          GUCAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUGUGUGCAUCUACUGCAGUGAGGGCACUUGUAGCAUUAUGCUGAC
          Chromosome Location Chr. 14 - 115043671 - 115043754 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000650
          miRBase ID: mmu-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAGGGCACUUGUAG
          Chromosome Location: Chr. 14 - 115043671 - 115043754 [+] on Build GRCm38

          miRBase Accession #: MIMAT0000649
          miRBase ID: mmu-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. 14 - 115043671 - 115043754 [+] on Build GRCm38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 143 bases
          Stem-loop ID mmu-mir-18a
          Stem-loop Accession # MI0000567
          Stem-loop Sequence
          UGCGUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGACUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUGUC
          Chromosome Location Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000528
          miRBase ID: mmu-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGAUAG
          Chromosome Location: Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004626
          miRBase ID: mmu-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 323 bases
          Stem-loop ID mmu-mir-19a
          Stem-loop Accession # MI0000688
          Stem-loop Sequence
          GCAGCCCUCUGUUAGUUUUGCAUAGUUGCACUACAAGAAGAAUGUAGUUGUGCAAAUCUAUGCAAAACUGAUGGUGGCCUGC
          Chromosome Location Chr. 14 - 115044000 - 115044081 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004660
          miRBase ID: mmu-miR-19a-5p
          Mature miRNA Sequence: UAGUUUUGCAUAGUUGCACUAC
          Chromosome Location: Chr. 14 - 115044000 - 115044081 [+] on Build GRCm38

          miRBase Accession #: MIMAT0000651
          miRBase ID: mmu-miR-19a-3p
          Mature miRNA Sequence: UGUGCAAAUCUAUGCAAAACUGA
          Chromosome Location: Chr. 14 - 115044000 - 115044081 [+] on Build GRCm38

          Gene Family ID MIPF0000011, mir-19
          Distance to Amplicon 472 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM13086
          Ambion® Pre-miR™ miRNA Precursor : PM13086
          TaqMan™ MicroRNA Assay : 002544
          mirVana® miRNA inhibitor : MH13086
          mirVana® miRNA mimic : MC13086
          TaqMan™ Advanced miRNA Assay : mmu481633_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12391
          Ambion® Pre-miR™ miRNA Precursor : PM12391
          TaqMan™ MicroRNA Assay : 002543
          mirVana® miRNA inhibitor : MH12391
          mirVana® miRNA mimic : MC12391
          TaqMan™ Advanced miRNA Assay : mmu481556_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12973
          Ambion® Pre-miR™ miRNA Precursor : PM12973
          TaqMan™ MicroRNA Assay : 002422
          mirVana® miRNA inhibitor : MH12973
          mirVana® miRNA mimic : MC12973
          TaqMan™ Advanced miRNA Assay : 478551_mir
          TaqMan™ Advanced miRNA Assay : rno480968_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10649
          Ambion® Pre-miR™ miRNA Precursor : PM10649
          TaqMan™ MicroRNA Assay : 000395
          mirVana® miRNA inhibitor : MH10649
          mirVana® miRNA mimic : MC10649
          TaqMan™ Advanced miRNA Assay : mmu479228_mir
          TaqMan™ Advanced miRNA Assay : rno479228_mir
          TaqMan™ Advanced miRNA Assay : 479228_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12283
          Ambion® Pre-miR™ miRNA Precursor : PM12283
          TaqMan™ MicroRNA Assay : 002490
          mirVana® miRNA inhibitor : MH12283
          mirVana® miRNA mimic : MC12283
          TaqMan™ Advanced miRNA Assay : mmu481574_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12412
          Ambion® Pre-miR™ miRNA Precursor : PM12412
          TaqMan™ MicroRNA Assay : 002308
          mirVana® miRNA inhibitor : MH12412
          mirVana® miRNA mimic : MC12412
          TaqMan™ Advanced miRNA Assay : 478447_mir


          miRBase Alias:

          Mouse: mmu-mir-18a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline