Search
Search
FIGURE: 1 / 98
Occludin is a 65 kDa protein that can exist in a variety of phosphorylated forms, ranging up to approximately 82 kDa. Occludin is thought to be involved in regulating both the localization and the function of occludin. Polyunsaturated fatty acids are known to up-regulate occludin expression, increasing the transendothelial cell resistance and reducing the cellular permeability to large molecules. The level of occludin varies greatly depending on tissue; in brain tissue, occludin is highly and continuously expressed at cell-cell contact sites, whereas non-neural tissues show lower expression and discontinuous distribution. Overall structural features of the occludin protein are highly conserved in all the species examined. Under-expression of tight junction proteins, including occludin, is a key molecular abnormality responsible for the increased permeability of tumor endothelial tight junctions, which contributes to brain tumor edemas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Occludin; phosphatase 1, regulatory subunit 115; tight junction protein; tight junction structural protein; unnamed protein product
Gene Aliases: AI503564; BLCPMG; Ocl; OCLN; PPP1R115; PTORCH1
UniProt ID: (Human) Q16625, (Dog) Q28269, (Mouse) Q61146, (Rat) Q6P6T5
Entrez Gene ID: (Human) 100506658, (Dog) 403844, (Mouse) 18260, (Rat) 83497
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support