Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • IL-13 Antibodies

          Invitrogen

          IL-13 Monoclonal Antibody (eBio13A), eFluor™ 660, eBioscience™

          Advanced Verification
          44 Published Figures
          46 References
          View all (87) IL-13 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL-13 Monoclonal Antibody (eBio13A), eFluor™ 660, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (44)
          • Advanced Verification (1)
          IL-13 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-13 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-13 Antibody (50-7133-82) in Flow

          BALB/c splenocytes were polarized under Th2 conditions for 3 days and then restimulated with Cell Stimulation Cocktail (plus protein transport inhibitors) for 5 hours. Following restimulation, cells were fixed and permeabilized and then intracellularly stained with Anti-Mouse CD4 FITC (Product # 11-0042-82) and 0.125 µg of Rat IgG1 K Isotype Control eFluor® 660 (Product # 50-4301-82) (left) or 0.125 µg of Anti-Mouse IL-13 eFluor® 660 (right). Total viable cells, as determined by Fixable Viability Dye eFluor® 450 (Product # 65-0863-14), wer... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry, ELISA (Flow, ELISA)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Neutralization (Neu)
          IL-13 Antibody

          Product Details

          50-7133-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.25 µg/test
          View 43 publications 43 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG1, kappa

          Recommended Isotype Control

          Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio13A

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660
          • Unconjugated
          • Alexa Fluor 488 
          • APC-eFluor 780
          • Brilliant UV 737
          • Brilliant UV 805
          • eFluor 450
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2574279

          Product Specific Information

          Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12)

          Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This eBio13A antibody has been tested by intracellular staining and flow cytometric analysis of Th2 polarized mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: ALRH; ALRH antibody; BHR1; il 13; IL-13; ILN; Interleukin; Interleukin-13; Interleukin13; MGC116786; MGC116788; MGC116789; NC30 antibody; P600 antibody; T-cell activation protein P600

          View more View less

          Gene Aliases: Il-13; Il13

          View more View less

          UniProt ID: (Mouse) P20109

          View more View less

          Entrez Gene ID: (Mouse) 16163

          View more View less

          Function(s)
          cytokine activity cytokine receptor binding interleukin-13 receptor binding
          Process(es)
          microglial cell activation positive regulation of immunoglobulin production inflammatory response immune response regulation of proton transport positive regulation of B cell proliferation positive regulation of connective tissue growth factor production negative regulation of NAD(P)H oxidase activity response to nicotine positive regulation of tyrosine phosphorylation of Stat6 protein positive regulation of macrophage activation positive regulation of ion transport positive regulation of mast cell degranulation positive regulation of smooth muscle cell proliferation positive regulation of protein secretion positive regulation of release of sequestered calcium ion into cytosol cellular response to cytokine stimulus negative regulation of transforming growth factor beta production negative regulation of neuron death negative regulation of complement-dependent cytotoxicity positive regulation of pancreatic stellate cell proliferation negative regulation of endothelial cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline