Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HuC/HuD Antibodies

          Invitrogen

          HuC/HuD Monoclonal Antibody (16A11)

          90 Published Figures
          692 References
          View all (3) HuC/HuD antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HuC/HuD Monoclonal Antibody (16A11)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          • Published Figures (90)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 97

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HuC/HuD Antibody (A-21271) in ICC/IF

          Immunofluorescent analysis of HuC/D (green) and MAP2 (red) on rat primary cortical neurons cultured for 28 days in the B-27 Plus Neuronal Culture System (Product # A3653401). At day 28 the cells were fixed with 4% paraformaldehyde for 15 min, permeabilized with 0.1% triton x-100 for 30min, and blocked with 1% BSA for 30 min at room temperature. Cells were stained with anti-HuC/D antibody (Product # A-21271) at a dilution of 1:250, and anti-MAP2 (Product # PA5-17646) at a dilution of 1:250, in 1% BSA staining buffer, overnight at 4C, and th... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunohistochemistry (Frozen) (IHC (F))
          HuC/HuD Antibody in Immunohistochemistry (Frozen) (IHC (F))
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          HuC/HuD Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunocytochemistry (ICC/IF)
          HuC/HuD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuC/HuD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuC/HuD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuC/HuD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuC/HuD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuC/HuD Antibody in Immunohistochemistry (Frozen) (IHC (F))
          HuC/HuD Antibody in Immunohistochemistry (Frozen) (IHC (F))
          HuC/HuD Antibody in Immunohistochemistry (Frozen) (IHC (F))
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry, In Situ Hybridization (IHC, ISH)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)
          HuC/HuD Antibody in Immunohistochemistry (IHC)

          Product Details

          A-21271

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          View 16 publications 16 publications

          Immunohistochemistry (IHC)

          -
          View 449 publications 449 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 28 publications 28 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          5-20 µg/mL
          View 54 publications 54 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 6 publications 6 publications

          Immunocytochemistry (ICC/IF)

          5-20 µg/mL
          View 90 publications 90 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 2 publications 2 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 41 publications 41 publications
          Product Specifications

          Species Reactivity

          Avian, Chicken, Human, Zebrafish

          Published species

          Amphibian, Avian, Bacteria, Cat, Chicken, Chimpanzee, Fish, Guinea pig, Horse, Human, Lizard, Mouse, Non-human primate, Pig, Rabbit, Rat, Reptile, Rhesus monkey, Rodent, Shark, Sheep, Shrew, Virus, Xenopus, Zebrafish

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          16A11

          Immunogen

          Human HuC/HuD neuronal protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • Request custom conjugation

          Form

          Lyophilized

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          HEPES buffered saline

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_221448

          Product Specific Information

          This antibody recognizes the Elav family members HuC, HuD and Hel-N1 neuronal proteins. It does not recognize HuR, another Elav family member that is present in all proliferating cells. The antibody has been shown to specifically label neuronal cells in zebrafish, chick, canaries, and humans, and is likely to label neuronal cells in most vertebrate species. Labeling is visible early in development, at about the time that the neurons leave the mitotic cycle.



          Storage and reconstitution: Upon receipt, lyophilized antibodies should be stored desiccated at -20°C or lower. When properly stored, these products are stable for at least one year. To prepare stock solution, reconstitute the antibody in 0.5-1 mL PBS, pH 7.4, containing 1% BSA. Store the solution for up to two weeks at 4°C with the addition of 2mM sodium azide. For longer storage, divide solutions into single-use aliquots and freeze at -20°C, avoidig freeze/thaw cycles.



          For best results in immunohistochemistry, antigen retrieval is required. Use 50mM Tris, pH 8.0, for 30 minutes.



          A-21271 was successfully used to detect HuC/HuD in neurons differentiated from H9 ESC derived NSCs.

          Target Information

          Anti-Hu antibodies were originally isolated from a patient with paraneoplastic encephalomyelitis, a rare condition of selective neural tissue injury associated with small cell lung carcinoma. The antibodies bind specifically to antigens present exclusively in neuronal cells and are thus useful as markers of neuronal cells in tissue. The Hu antigen is an RNA-binding protein of the embryonic lethal abnormal visual (Elav) family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 3; ELAV like neuron-specific RNA binding protein 4; ELAV-like protein 3; ELAV-like protein 4; elav-related C; elav-related D; elavC; embryonic lethal, abnormal vision-related C; embryonic lethal, abnormal vision-related D; etID309927.3; Hu antigen C; Hu antigen D; Hu-antigen C; Hu-antigen D; HuC; HuD; Paraneoplastic cerebellar degeneration-associated antigen; paraneoplastic encephalomyelitis antigen; Paraneoplastic encephalomyelitis antigen HuD; Paraneoplastic limbic encephalitis antigen 21; Protein elrD; RNA-binding protein HuD; unnamed protein product

          View more View less

          Gene Aliases: ELAVL3; ELAVL4; elrc; elrd; HUC; HUCL; HUD; id:ibd1248; PLE21; PNEM; wu:fb77b03; wu:fc24h01; zHuC

          View more View less

          UniProt ID: (Human) Q14576, (Human) P26378, (Zebrafish) A0A0R4IEW8

          View more View less

          Entrez Gene ID: (Human) 1995, (Chicken) 395634, (Human) 1996, (Zebrafish) 30732, (Zebrafish) 30737

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA 3'-UTR binding protein binding mRNA 3'-UTR AU-rich region binding mRNA binding poly(A) binding pre-mRNA intronic pyrimidine-rich binding translation initiation factor
          Process(es)
          nervous system development cell differentiation RNA processing mRNA processing learning locomotory behavior RNA splicing cerebral cortex neuron differentiation dendrite morphogenesis 3'-UTR-mediated mRNA stabilization positive regulation of 3'-UTR-mediated mRNA stabilization regulation of neurogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline