Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
Positive Control: Rat cerebellum tissue lysate, SK-Br-3 cell lysate, A549 cell lysate, mouse cerebellum tissue lysate, F9, human liver carcinoma tissue, human cervix tissue, human colon carcinoma tissue, human breast carcinoma tissue, human esophagus tissue, human stomach carcinoma tissue, human small intestine tissue, THP-1.
Subcellular Location: Nucleus, Cytoplasm, Extracellular exosome.
HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686J1764; epididymis secretory sperm binding protein; FLJ96580; HDGF; hepatoma derived growth factor; Hepatoma-derived growth factor; High mobility group protein 1-like 2; HMG-1L2; unnamed protein product
Gene Aliases: AI118077; D3Ertd299e; HDGF; HMG1L2; Tdrm1
UniProt ID: (Human) P51858, (Rat) Q8VHK7, (Mouse) P51859
Entrez Gene ID: (Human) 3068, (Rat) 114499, (Mouse) 15191
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support