Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Pan SHANK Antibodies

          Invitrogen

          Pan SHANK Monoclonal Antibody (N23b/49), PE

          View all (12) Pan SHANK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Pan SHANK Monoclonal Antibody (N23b/49), PE

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Pan SHANK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Pan SHANK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Pan SHANK Antibody (MA5-45441) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using human neuroblastoma cells. Fixation involved 4% Formaldehyde for 15 min at RT. Samples were incubated with SHANK (pan) monoclonal antibody (Product # MA5-45441) at 1:100 for 60 min at RT, followed by Goat Anti-Mouse ATTO 488 at 1:200 for 60 min at RT. Counterstain used was Phalloidin Texas Red F-Actin stain; DAPI (blue) nuclear stain at 1:1,000, 1:5,000 ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Pan SHANK Antibody in Immunocytochemistry (ICC/IF)
          Pan SHANK Antibody in Immunocytochemistry (ICC/IF)
          Pan SHANK Antibody in Western Blot (WB)
          Pan SHANK Monoclonal Antibody (N23b/49), PE

          Product Details

          MA5-45441

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Antibody Microarray (Ab Microarray)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N23b/49

          Immunogen

          Fusion protein amino acids 84-309 of rat Shank2
          3D Epitope / Immunogen

          Conjugate

          PE PE PE
          • Unconjugated
          • APC 
          • FITC
          • PerCP 
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2931895

          Product Specific Information

          1 µg/mL of MA5-45441 was sufficient for detection of Shank1-4 in 10 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 160kDa. Recognizes Shank1, 2 and 3.

          Target Information

          Shank proteins make up a family of scaffold proteins identified through their interaction with a variety of membrane and cytoplasmic proteins. Shank proteins at postsynaptic sites of excitatory synapses play roles in signal transmission into the postsynaptic neuron. Studies suggest that Shank2 is expressed in the neurons of the developing retina, and could play a role in the neuronal differentiation of the developing retina. Other recent studies suggest that the disruption of glutamate receptors at the Shank postsynaptic platform could contribute to the destruction of the postsynaptic density, which underlies the synaptic dysfunction and loss in Alzheimer's disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cortactin SH3 domain-binding protein; Cortactin-binding protein 1; CortBP1; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; multidomain postsynaptic density protein; PDZ-based scaffold protein; ProSAP1 protein; postsynaptic scaffold protein; proline rich synapse associated protein 1; proline rich synapse associated protein 2; proline-rich synapse associated protein 1; Proline-rich synapse-associated protein 1; Proline-rich synapse-associated protein 2; ProSAP1; ProSAP2; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3 and multiple ankyrin repeat domains protein 2; SH3 and multiple ankyrin repeat domains protein 3; SH3 and multiple ankyrin repeat domains protein 3 variant c-3; SH3 and multiple ankyrin repeat domains protein 3 variant c-4; SH3 and multiple ankyrin repeat domains-like protein 3 variant 1; SH3 and multiple ankyrin repeat domains-like protein 3 variant 2; SH3 and multiple ankyrin repeat domains-like protein 3 variant 3; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-3; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-2; SH3/ankyrin domain gene 1; SH3/ankyrin domain gene 2; SH3/ankyrin domain gene 3; shank postsynaptic density protein; Shank postsynaptic density protein 3a; Shank1; Shank2; Shank3; shank3 postsynaptic density protein; shank3 splice variant II; shank3 splice variant III; shank3 splice variant IV; shank3e splice variant III; shank3e splice variant IV; Somatostatin receptor-interacting protein; SPANK-1; SPANK-2; SPANK-3; SSTR-interacting protein; Synamon

          View more View less

          Gene Aliases: AI841104; AUTS17; CORTBP1; CTTNBP1; DEL22q13.3; KIAA1022; KIAA1650; mKIAA1022; PROSAP1; PROSAP2; PSAP2; SCZD15; SHANK; SHANK1; Shank1a; SHANK2; SHANK3; Shank3b; SHANK3c-3; SHANK3c-4; SPANK-1; SPANK-2; SPANK-3; Spank1; SSTRIP; synamon

          View more View less

          UniProt ID: (Human) Q9Y566, (Human) Q9UPX8, (Human) Q9BYB0, (Rat) Q9WV48, (Mouse) D3YZU1, (Mouse) Q80Z38, (Rat) Q9QX74, (Mouse) Q4ACU6, (Rat) Q9JLU4

          View more View less

          Entrez Gene ID: (Human) 50944, (Human) 22941, (Human) 85358, (Rat) 78957, (Mouse) 243961, (Mouse) 210274, (Rat) 171093, (Mouse) 58234, (Rat) 59312

          View more View less

          Function(s)
          G-protein coupled receptor binding protein binding SH3 domain binding receptor signaling complex scaffold activity GKAP/Homer scaffold activity somatostatin receptor binding ionotropic glutamate receptor binding identical protein binding macromolecular complex binding ankyrin repeat binding scaffold protein binding structural constituent of postsynaptic density actin binding zinc ion binding protein C-terminus binding protein complex binding protein self-association
          Process(es)
          MAPK cascade embryonic epithelial tube formation synapse assembly learning memory striatal medium spiny neuron differentiation adult behavior negative regulation of actin filament bundle assembly social behavior locomotory exploration behavior vocal learning negative regulation of cell volume positive regulation of long-term neuronal synaptic plasticity brain morphogenesis neuromuscular process controlling balance protein oligomerization positive regulation of synapse structural plasticity positive regulation of synaptic transmission, glutamatergic dendritic spine morphogenesis positive regulation of dendritic spine development regulation of dendritic spine morphogenesis vocalization behavior postsynaptic density assembly AMPA glutamate receptor clustering NMDA glutamate receptor clustering guanylate kinase-associated protein clustering regulation of long-term synaptic potentiation positive regulation of long-term synaptic potentiation positive regulation of glutamate receptor signaling pathway regulation of long term synaptic depression positive regulation of excitatory postsynaptic potential regulation of grooming behavior regulation of behavioral fear response positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity associative learning synapse organization cognition exploration behavior long-term synaptic potentiation long term synaptic depression nervous system development long-term memory cell differentiation protein localization to synapse olfactory behavior habituation modulation of synaptic transmission determination of affect righting reflex synapse maturation protein-containing complex assembly AMPA selective glutamate receptor signaling pathway maintenance of postsynaptic density structure positive regulation of cell proliferation negative regulation of hippo signaling protein complex assembly cytoskeletal anchoring at plasma membrane regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity brain development response to nutrient
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline