Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GluR1 Antibodies

          Invitrogen

          GluR1 Monoclonal Antibody (N355/1), PE

          View all (48) GluR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GluR1 Monoclonal Antibody (N355/1), PE

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          GluR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GluR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GluR1 Antibody (MA5-45630) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using human neuroblastoma cells. Fixation involved 4% Formaldehyde for 15 min at RT. Samples were incubated with GluA1/GluR1 Glutamate Receptor monoclonal antibody (Product # MA5-45630) at 1:100 for 60 min at RT, followed by Goat Anti-Mouse ATTO 488 at 1:100 for 60 min at RT. Counterstain used was Phalloidin Texas Red F-Actin stain; DAPI (blue) nuclear stain at 1:1,000, 1:5,000 for 60 min RT, 5 min RT. Localization: Cell Membrane, Cell Junction. Magnification: 60X. (A) DAPI (blue) nuclear sta... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GluR1 Antibody in Immunocytochemistry (ICC/IF)
          GluR1 Antibody in Immunocytochemistry (ICC/IF)
          GluR1 Antibody in Western Blot (WB)
          GluR1 Monoclonal Antibody (N355/1), PE

          Product Details

          MA5-45630

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N355/1

          Immunogen

          Fusion protein amino acids 1-389 (extracellular N-terminus) of rat GluA1/GluR1
          3D Epitope / Immunogen

          Conjugate

          PE PE PE
          • Unconjugated
          • APC 
          • FITC
          • PerCP 
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          95.46mM phosphate/2.48mM MES, pH 7.4, with 2mM EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2932084

          Product Specific Information

          1 µg/mL of MA5-45630 was sufficient for detection of GluA1/GluR1 in 20 µg of mouse brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 100kDa. Does not cross-react with GluR2.

          This antibody was formerly sold as clone S355-1.

          Target Information

          GluR1 (glutamate receptor 1) is an ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter, L-glutamate, induces a conformation change that leads to the opening of the cation channel and converts the chemical signal to an electrical impulse. Then, the receptor desensitizes rapidly and enters a transiet inactive state characterized by the presence of a bound agonist. In the presence of CACNG4, CACNG7, or CACNG8 GluR1 shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AI853806; AMPA 1; AMPA receptor subunit GluA1; AMPA-selective glutamate receptor 1; GluA1; GLUH 1; GluR K1; GluR-1; GluR-A; GluR-K1; GluR1; GluRK1; glutamate receptor; Glutamate receptor 1; glutamate receptor A; Glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1); GPRC1A; Gria 1; GRM1A; HGBR1; HIPA; MGC13325; MGC133252; NMDA1; NMDAR1; NR1; OTTHUMP00000160643; OTTHUMP00000165781; OTTHUMP00000224241; OTTHUMP00000224242; OTTHUMP00000224243; RP11-350O14.1; unnamed protein product

          View more View less

          Gene Aliases: 2900051M01Rik; Glr-1; Glr1; GLUA1; GLUH1; Glur-1; GluR-A; gluR-K1; GLUR1; GLURA; GRIA1; HBGR1; HIPA1; MRD67; MRT76

          View more View less

          UniProt ID: (Human) P42261, (Rat) P19490, (Mouse) P23818

          View more View less

          Entrez Gene ID: (Human) 2890, (Rat) 50592, (Mouse) 14799

          View more View less

          Function(s)
          beta-amyloid binding G-protein alpha-subunit binding ionotropic glutamate receptor activity AMPA glutamate receptor activity ion channel activity protein binding adenylate cyclase binding ligand-gated ion channel activity immunoglobulin binding protein kinase binding protein domain specific binding glutamate-gated calcium ion channel activity PDZ domain binding small GTPase binding myosin V binding G-protein beta-subunit binding beta-2 adrenergic receptor binding glutamate receptor binding signaling receptor activity identical protein binding protein kinase A binding peptide hormone receptor binding scaffold protein binding ligand-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential extracellular-glutamate-gated ion channel activity protein homodimerization activity receptor activity transmembrane signal receptor
          Process(es)
          regulation of receptor recycling ion transport signal transduction chemical synaptic transmission synapse assembly long-term memory response to xenobiotic stimulus response to toxic substance response to sucrose response to lithium ion positive regulation of gene expression neuronal action potential calcium-mediated signaling spinal cord development cerebral cortex development receptor internalization response to nutrient levels response to estradiol ion transmembrane transport regulation of ion transmembrane transport ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic response to cocaine response to morphine response to peptide hormone response to ethanol positive regulation of membrane potential response to arsenic-containing substance regulation of synaptic plasticity behavioral response to pain modulation of synaptic transmission positive regulation of synaptic transmission response to electrical stimulus regulation of postsynaptic membrane potential long-term synaptic potentiation long term synaptic depression response to fungicide cellular response to amino acid stimulus cellular response to ammonium ion cellular response to dsRNA cellular response to growth factor stimulus cellular response to peptide hormone stimulus cellular response to amine stimulus positive regulation of locomotion involved in locomotory behavior regulation of presynaptic membrane potential cellular response to L-glutamate cellular response to brain-derived neurotrophic factor stimulus conditioned place preference response to psychosocial stress positive regulation of excitatory postsynaptic potential response to organic cyclic compound response to drug cellular response to organic cyclic compound transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline