Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Cochlin is a secreted protein encoded by the coagulation factor C homology (COCH) gene, a cochlear gene. It constitutes 70% of the inner ear proteins and is classified into three glycosylated isoforms: p63s, p44s and p40. Cochlin contains an N-terminal LCCL domain and two von Willebrand factor A-like domains. Mutations in the COCH gene cause DFNA9, an autosomal dominant nonsyndromic auditory and vestibular dysfunction disorder, as a result of either an amino acid deletion in the LCCL domain or missense substitutions. Microfibrillar deposits accumulate in the inner ear of individuals with DFNA9 and these deposits may contain the Cochlin protein. Cochlin is a target antigen for autoimmune sensorineural hearing loss.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coagulation factor C homolog, cochlin (Limulus polyphemus); COCH-5B2; Cochlin; PRO294; unnamed protein product; UNQ257
Gene Aliases: COCH; COCH-5B2; COCH5B2; DFNA9; DFNB110; UNQ257/PRO294
UniProt ID: (Human) O43405
Entrez Gene ID: (Human) 1690
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support