Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ASCL1
Transcript:
NM_004316.3
Assay Name:
Mutation genome location:
chr.12 102958246-102958749 on build GRCh38
Assay transcription exon location:
Within Exon 1
Assay ID
Hs00126424_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neuron migration
noradrenergic neuron development
noradrenergic neuron fate commitment
transcription, DNA-templated
Notch signaling pathway
regulation of mitotic cell cycle
neuroblast fate determination
neuroblast proliferation
response to lithium ion
regulation of gene expression
oligodendrocyte development
spinal cord association neuron differentiation
spinal cord oligodendrocyte cell fate specification
vestibular nucleus development
cerebral cortex GABAergic interneuron differentiation
commitment of neuronal cell to specific neuron type in forebrain
central nervous system neuron development
cerebral cortex development
neurogenesis
regulation of epithelial cell differentiation
response to retinoic acid
negative regulation of apoptotic process
positive regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of Notch signaling pathway
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
sympathetic nervous system development
neuron fate specification
musculoskeletal movement, spinal reflex action
response to folic acid
subpallium neuron fate commitment
regulation of timing of subpallium neuron differentiation
olfactory pit development
lung epithelial cell differentiation
ventral spinal cord interneuron fate commitment
lung neuroendocrine cell differentiation
stomach neuroendocrine cell differentiation
carotid body glomus cell differentiation
adrenal chromaffin cell differentiation
sympathetic ganglion development
response to epidermal growth factor
cellular response to magnetism
positive regulation of neural precursor cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
bHLH transcription factor binding
E-box binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline