Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
AUP1
Transcript:
NM_181575.4
Assay Name:
Mutation genome location:
chr.2 74529263-74529753 on build GRCh38
Assay transcription exon location:
Overlaps Exon 1 - Exon 3
Gene:
HTRA2
Transcript:
NM_181575.4
Assay Name:
Mutation genome location:
chr.2 74529263-74529753 on build GRCh38
Assay transcription exon location:
Overlaps Exon 1 - Exon 3
Assay ID
Hs00195699_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
scaffold/adaptor protein
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
ER-associated ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
regulation of catalytic activity
proteolysis
ceramide metabolic process
mitochondrion organization
adult walking behavior
intrinsic apoptotic signaling pathway in response to DNA damage
response to herbicide
positive regulation of cell death
protein autoprocessing
pentacyclic triterpenoid metabolic process
forebrain development
cellular response to oxidative stress
cellular response to heat
cellular response to interferon-beta
regulation of multicellular organism growth
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
cellular protein catabolic process
negative regulation of cell cycle
neuron development
cellular response to retinoic acid
cellular response to growth factor stimulus
execution phase of apoptosis
negative regulation of neuron death
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
protein binding
ubiquitin binding
ubiquitin-protein transferase activator activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
unfolded protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline