Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CD14
Transcript:
NM_000591.3
Assay Name:
Mutation genome location:
chr.5 140631589-140632094 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 2
Gene:
TMCO6
Transcript:
NM_000591.3
Assay Name:
Mutation genome location:
chr.5 140631589-140632094 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 2
Assay ID
Hs00266191_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
MyD88-independent toll-like receptor signaling pathway
receptor-mediated endocytosis
phagocytosis
apoptotic process
inflammatory response
cell surface receptor signaling pathway
I-kappaB kinase/NF-kappaB signaling
response to heat
lipopolysaccharide-mediated signaling pathway
response to magnesium ion
positive regulation of type I interferon production
positive regulation of interferon-gamma production
positive regulation of tumor necrosis factor production
negative regulation of MyD88-independent toll-like receptor signaling pathway
toll-like receptor 4 signaling pathway
response to tumor necrosis factor
TRIF-dependent toll-like receptor signaling pathway
toll-like receptor TLR1:TLR2 signaling pathway
toll-like receptor TLR6:TLR2 signaling pathway
innate immune response
response to ethanol
positive regulation of endocytosis
positive regulation of cytokine secretion
response to electrical stimulus
necroptotic process
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
cellular response to diacyl bacterial lipopeptide
cellular response to triacyl bacterial lipopeptide
apoptotic signaling pathway
protein import into nucleus
lipopolysaccharide binding
opsonin receptor activity
protein binding
peptidoglycan receptor activity
lipoteichoic acid binding
protein transporter activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline