Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
GATA3
Transcript:
NM_001002295.1
Assay Name:
Mutation genome location:
chr.10 8069391-8069889 on build GRCh38
Assay ID
Hs00316463_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
cell fate determination
neuron migration
type IV hypersensitivity
kidney development
mesonephros development
lens development in camera-type eye
pro-T cell differentiation
aortic valve morphogenesis
cardiac right ventricle morphogenesis
ventricular septum development
chromatin remodeling
transcription from RNA polymerase II promoter
defense response
humoral immune response
signal transduction
axon guidance
heart development
blood coagulation
negative regulation of cell proliferation
male gonad development
response to virus
anatomical structure morphogenesis
post-embryonic development
organ morphogenesis
positive regulation of signal transduction
response to gamma radiation
positive regulation of endothelial cell migration
regulation of neuron projection development
phosphatidylinositol 3-kinase signaling
erythrocyte differentiation
TOR signaling
negative regulation of interferon-gamma production
negative regulation of interleukin-2 production
positive regulation of interleukin-4 production
negative regulation of mammary gland epithelial cell proliferation
embryonic hemopoiesis
cellular response to interferon-alpha
ureter maturation
parathyroid hormone secretion
regulation of cytokine biosynthetic process
norepinephrine biosynthetic process
inner ear morphogenesis
response to drug
regulation of CD4-positive, alpha-beta T cell differentiation
regulation of neuron apoptotic process
ear development
response to estrogen
thymic T cell selection
T-helper 2 cell differentiation
innate immune response
cell fate commitment
response to ethanol
positive regulation of T cell differentiation
negative regulation of fat cell differentiation
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
sympathetic nervous system development
thymus development
digestive tract development
developmental growth
anatomical structure formation involved in morphogenesis
negative regulation of inflammatory response
T cell receptor signaling pathway
regulation of histone H3-K4 methylation
positive regulation of protein kinase B signaling
parathyroid gland development
pharyngeal system development
uterus development
mesenchymal to epithelial transition
mast cell differentiation
ureteric bud formation
regulation of histone H3-K27 methylation
canonical Wnt signaling pathway involved in metanephric kidney development
cellular response to interleukin-4
cellular response to tumor necrosis factor
positive regulation of histone H3-K14 acetylation
otic vesicle development
cellular response to BMP stimulus
positive regulation of ureteric bud formation
nephric duct morphogenesis
nephric duct formation
regulation of nephron tubule epithelial cell differentiation
interleukin-4 secretion
interferon-gamma secretion
lymphocyte migration
negative regulation of DNA demethylation
regulation of establishment of cell polarity
negative regulation of cell motility
negative regulation of endothelial cell apoptotic process
positive regulation of T-helper 2 cell cytokine production
negative regulation of cell proliferation involved in mesonephros development
positive regulation of thyroid hormone generation
positive regulation of histone H3-K9 acetylation
positive regulation of interleukin-5 secretion
positive regulation of interleukin-13 secretion
positive regulation of transcription regulatory region DNA binding
regulation of cellular response to X-ray
negative regulation of fibroblast growth factor receptor signaling pathway involved in ureteric bud formation
negative regulation of glial cell-derived neurotrophic factor receptor signaling pathway involved in ureteric bud formation
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
nucleic acid binding transcription factor activity
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
interleukin-2 receptor binding
protein binding
transcription factor binding
zinc ion binding
transcription regulatory region DNA binding
protein dimerization activity
E-box binding
HMG box domain binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline