Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CDK5R1
Transcript:
NM_003885.2
Assay Name:
Mutation genome location:
chr.17 32487897-32488393 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00347575_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
kinase activator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
neuron migration
neuron cell-cell adhesion
G-protein coupled acetylcholine receptor signaling pathway
axon guidance
axonal fasciculation
brain development
cell proliferation
embryo development
regulation of macroautophagy
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cerebellum development
superior olivary nucleus maturation
hippocampus development
layer formation in cerebral cortex
neuron differentiation
negative regulation of axon extension
neuron projection development
regulation of actin cytoskeleton organization
serine phosphorylation of STAT3 protein
ionotropic glutamate receptor signaling pathway
positive regulation of neuron apoptotic process
regulation of neuron differentiation
negative regulation of transcription, DNA-templated
ephrin receptor signaling pathway
rhythmic process
regulation of dendritic spine morphogenesis
regulation of microtubule cytoskeleton organization
positive regulation of cell cycle arrest
positive regulation of protein serine/threonine kinase activity
positive regulation of protein targeting to membrane
regulation of signal transduction by p53 class mediator
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
calcium ion binding
protein binding
kinase activity
cyclin-dependent protein kinase 5 activator activity
protein kinase binding
protein serine/threonine kinase activator activity
cadherin binding
ephrin receptor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline