Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
PEX11B
Entrez Gene ID
8799
Gene Name:
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.714608
Species:
Human
Primer Chromosome Location
Chr.1: 145912669-145912132 on Build GRCh38
View in UCSC browser
Amplicon Length
538
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00399859_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
PEX11B
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001184795.1
NP_001171724.1
-
-
-
NM_003846.2
NP_003837.1
6744090
c.669C>A
p.D223E
3862586
c.725C>T
p.S242F
6744083
c.427C>T
p.R143C
3801644
c.393C>T
p.L131L
7126548
c.596G>C
p.R199P
423695
c.720C>T
p.L240L
423694
c.436A>G
p.M146V
7096449
c.588C>G
p.L196L
674808
c.634G>C
p.V212L
4142532
c.450T>A
p.S150S
6744092
c.693C>T
p.G231G
4021752
c.672A>G
p.K224K
6009442
c.430C>T
p.L144L
283963
c.724T>C
p.S242P
5843456
c.595C>T
p.R199*
6574531
c.505G>C
p.E169Q
3788599
c.452C>T
p.S151F
3473100
c.757C>T
p.P253S
5461974
c.466C>T
p.R156*
1688458
c.493_494CC>GT
p.P165V
895186
c.649G>A
p.D217N
5377496
c.657C>T
p.F219F
895185
c.630C>T
p.D210D
5641208
c.420T>A
p.Y140*
1731859
c.375-3C>G
p.?
5625879
c.758C>T
p.P253L
5521307
c.439G>A
p.E147K
1725640
c.397A>G
p.M133V
4576222
c.729C>T
p.S243S
1197580
c.467G>C
p.R156P
4504040
c.654C>T
p.L218L
4495236
c.449C>T
p.S150F
7135131
c.399G>T
p.M133I
5117318
c.649G>T
p.D217Y
895184
c.589C>A
p.L197M
1600713
c.750A>G
p.L250L
5052498
c.766C>T
p.R256*
895183
c.409C>T
p.R137C
4766606
c.683G>A
p.W228*
1491660
c.408C>T
p.S136S
1287079
c.774G>T
p.K258N
4713653
c.478T>C
p.S160P
NR_073491.1
-
-
-
NR_073492.1
-
-
-
NR_073493.2
-
-
-
GenBank
AB018080.1
BAA34812.1
-
-
-
AF093670.1
AAC78660.1
-
-
-
AK023991.1
-
-
-
AK094173.1
-
-
-
AK301983.1
-
-
-
BC011963.1
AAH11963.1
-
-
-
CB155824.1
-
-
-
CR542047.1
CAG46844.1
-
-
-
ENST00000369306.0
-
-
-
HY108328.1
-
-
-
Phenotype
MIM: 603867
Literature Links:
PEX11B
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
peroxisome organization
signal transduction
peroxisome fission
regulation of peroxisome size
protein homooligomerization
protein binding
protein homodimerization activity
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-vvmmd:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0