Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
APOE
Transcript:
NM_001302691.1
Assay Name:
Mutation genome location:
chr.19 44908802-44909368 on build GRCh38
Assay transcription exon location:
Within Exon 4
Assay ID
Hs00404452_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
retinoid metabolic process
negative regulation of endothelial cell proliferation
response to dietary excess
triglyceride metabolic process
cholesterol catabolic process
cellular calcium ion homeostasis
receptor-mediated endocytosis
cytoskeleton organization
G-protein coupled receptor signaling pathway
nitric oxide mediated signal transduction
synaptic transmission, cholinergic
cholesterol metabolic process
regulation of gene expression
negative regulation of platelet activation
positive regulation of cholesterol esterification
positive regulation of cholesterol efflux
long-chain fatty acid transport
protein import
virion assembly
triglyceride catabolic process
cGMP-mediated signaling
negative regulation of blood coagulation
regulation of axon extension
positive regulation of cGMP biosynthetic process
neuron projection regeneration
regulation of Cdc42 protein signal transduction
positive regulation of low-density lipoprotein particle receptor catabolic process
cholesterol efflux
phospholipid efflux
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
chylomicron remnant clearance
high-density lipoprotein particle clearance
very-low-density lipoprotein particle clearance
lipoprotein metabolic process
lipoprotein biosynthetic process
lipoprotein catabolic process
vasodilation
cholesterol homeostasis
negative regulation of MAP kinase activity
negative regulation of neuron apoptotic process
negative regulation of blood vessel endothelial cell migration
reverse cholesterol transport
positive regulation by host of viral process
negative regulation of cholesterol biosynthetic process
positive regulation of lipid biosynthetic process
intracellular transport
regulation of neuronal synaptic plasticity
artery morphogenesis
negative regulation of inflammatory response
positive regulation of nitric-oxide synthase activity
positive regulation of membrane protein ectodomain proteolysis
maintenance of location in cell
fatty acid homeostasis
positive regulation of dendritic spine development
negative regulation of canonical Wnt signaling pathway
AMPA glutamate receptor clustering
NMDA glutamate receptor clustering
cellular oxidant detoxification
regulation of beta-amyloid clearance
negative regulation of neuron death
positive regulation of postsynaptic membrane organization
negative regulation of presynaptic membrane organization
negative regulation of beta-amyloid formation
positive regulation of dendritic spine maintenance
positive regulation of phospholipid efflux
positive regulation of lipid transport across blood brain barrier
beta-amyloid binding
lipid transporter activity
protein binding
phospholipid binding
heparin binding
lipid binding
cholesterol binding
antioxidant activity
cholesterol transporter activity
identical protein binding
protein homodimerization activity
metal chelating activity
tau protein binding
low-density lipoprotein particle receptor binding
phosphatidylcholine-sterol O-acyltransferase activator activity
very-low-density lipoprotein particle receptor binding
lipoprotein particle binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline