Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HFE
Transcript:
NM_000410.3
Assay Name:
Mutation genome location:
chr.6 26092474-26093055 on build GRCh38
Assay ID
Hs00419627_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
major histocompatibility complex protein
Gene Ontology Categories:
Function(s)
Process(es)
antigen processing and presentation of peptide antigen via MHC class I
negative regulation of T cell antigen processing and presentation
negative regulation of T cell cytokine production
protein complex assembly
cellular iron ion homeostasis
acute-phase response
immune response
female pregnancy
response to iron ion
cellular response to iron ion starvation
positive regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
antigen processing and presentation
BMP signaling pathway
positive regulation of protein binding
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
hormone biosynthetic process
positive regulation of receptor-mediated endocytosis
iron ion homeostasis
multicellular organismal iron ion homeostasis
cellular response to iron ion
positive regulation of peptide hormone secretion
liver regeneration
iron ion import into cell
negative regulation of receptor binding
positive regulation of receptor binding
positive regulation of ferrous iron import into cell
negative regulation of antigen processing and presentation of endogenous peptide antigen via MHC class I
positive regulation of ferrous iron binding
positive regulation of transferrin receptor binding
response to iron ion starvation
regulation of protein localization to cell surface
negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of receptor activity
positive regulation of receptor activity
negative regulation of CD8-positive, alpha-beta T cell activation
antigen binding
receptor binding
protein binding
beta-2-microglobulin binding
co-receptor binding
peptide antigen binding
transferrin receptor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline