Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CDK5
Transcript:
NM_004935.3
Assay Name:
Mutation genome location:
chr.7 151056514-151057111 on build GRCh38
Assay ID
Hs00427855_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
synaptic transmission, dopaminergic
intracellular protein transport
nucleocytoplasmic transport
cell cycle
cell-matrix adhesion
chemical synaptic transmission
synapse assembly
skeletal muscle tissue development
motor neuron axon guidance
cell proliferation
visual learning
response to wounding
Schwann cell development
synaptic vesicle exocytosis
regulation of macroautophagy
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
sensory perception of pain
cerebellar cortex formation
hippocampus development
layer formation in cerebral cortex
central nervous system neuron development
corpus callosum development
neuron differentiation
regulation of cell migration
negative regulation of axon extension
cortical actin cytoskeleton organization
neuron projection development
negative regulation of protein ubiquitination
negative regulation of synaptic plasticity
positive regulation of protein binding
receptor catabolic process
serine phosphorylation of STAT3 protein
synaptic transmission, glutamatergic
protein localization to synapse
regulation of apoptotic process
receptor clustering
positive regulation of neuron apoptotic process
negative regulation of cell cycle
positive regulation of protein kinase activity
negative regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of calcium ion-dependent exocytosis
protein autophosphorylation
negative regulation of protein export from nucleus
behavioral response to cocaine
regulation of synaptic plasticity
synaptic vesicle endocytosis
rhythmic process
axon extension
oligodendrocyte differentiation
dendrite morphogenesis
cell division
neuron apoptotic process
excitatory postsynaptic potential
regulation of dendritic spine morphogenesis
calcium ion import
regulation of cell cycle arrest
positive regulation of protein targeting to membrane
negative regulation of neuron death
regulation of signal transduction by p53 class mediator
regulation of synaptic vesicle recycling
positive regulation of actin cytoskeleton reorganization
positive regulation of receptor activity
p53 binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ErbB-2 class receptor binding
protein binding
ATP binding
cytoskeletal protein binding
kinase activity
protein kinase binding
acetylcholine receptor activator activity
ErbB-3 class receptor binding
ephrin receptor binding
tau-protein kinase activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline