Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
BCL2
Transcript:
Assay Name:
Mutation genome location:
chr.18 63124479-63125072 on build GRCh38
Assay ID
Hs00432732_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
ossification
ovarian follicle development
metanephros development
branching involved in ureteric bud morphogenesis
behavioral fear response
B cell homeostasis
release of cytochrome c from mitochondria
regulation of cell-matrix adhesion
lymphoid progenitor cell differentiation
B cell lineage commitment
response to ischemia
renal system process
protein dephosphorylation
melanin metabolic process
regulation of nitrogen utilization
apoptotic process
humoral immune response
cellular response to DNA damage stimulus
actin filament organization
axonogenesis
female pregnancy
cell aging
male gonad development
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
intrinsic apoptotic signaling pathway in response to oxidative stress
response to radiation
response to toxic substance
post-embryonic development
response to iron ion
response to UV-B
response to gamma radiation
regulation of gene expression
negative regulation of autophagy
negative regulation of calcium ion transport into cytosol
regulation of glycoprotein biosynthetic process
mesenchymal cell development
positive regulation of neuron maturation
positive regulation of smooth muscle cell migration
cell growth
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cochlear nucleus development
gland morphogenesis
regulation of transmembrane transporter activity
negative regulation of ossification
positive regulation of cell growth
negative regulation of cell growth
melanocyte differentiation
negative regulation of cell migration
positive regulation of B cell proliferation
hair follicle morphogenesis
axon regeneration
regulation of protein stability
endoplasmic reticulum calcium ion homeostasis
glomerulus development
negative regulation of cellular pH reduction
negative regulation of myeloid cell apoptotic process
T cell differentiation in thymus
positive regulation of peptidyl-serine phosphorylation
negative regulation of osteoblast proliferation
response to cytokine
response to nicotine
organ growth
positive regulation of multicellular organism growth
B cell proliferation
cellular response to glucose starvation
response to drug
response to hydrogen peroxide
T cell homeostasis
negative regulation of apoptotic process
positive regulation of catalytic activity
CD8-positive, alpha-beta T cell lineage commitment
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
negative regulation of neuron apoptotic process
ear development
regulation of viral genome replication
positive regulation of melanocyte differentiation
negative regulation of retinal cell programmed cell death
regulation of mitochondrial membrane permeability
focal adhesion assembly
spleen development
thymus development
digestive tract morphogenesis
oocyte development
positive regulation of skeletal muscle fiber development
pigment granule organization
homeostasis of number of cells within a tissue
B cell receptor signaling pathway
response to glucocorticoid
neuron apoptotic process
defense response to virus
regulation of mitochondrial membrane potential
negative regulation of mitochondrial depolarization
regulation of calcium ion transport
transmembrane transport
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to organic substance
cellular response to hypoxia
reactive oxygen species metabolic process
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
negative regulation of G1/S transition of mitotic cell cycle
negative regulation of reactive oxygen species metabolic process
negative regulation of anoikis
negative regulation of apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
positive regulation of intrinsic apoptotic signaling pathway
protease binding
protein binding
transcription factor binding
channel activity
channel inhibitor activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity
BH3 domain binding
protein phosphatase 2A binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-2026.03.11-1.0